Learn More
Gibco™ Hepatic Spheroid Kit
Description
The Gibco™ Hepatic Spheroid Kit is the first off-the-shelf kit for generation of hepatic spheroids in your own lab.
- Maintain cells in culture longer – up to 21 days, >4X longer than cells last in monolayer culture
- Design your own schedule – cells are ready for plating when you are
- Interrogate pathways of interest – applicable for toxicity, metabolism, or disease modeling
What you get:
- Spheroid-qualified hepatocytes
- Hepatocyte Thaw Medium
- Plating supplement (Cocktail A + Dexamethasone)
- Maintenance supplement (Cocktail B + Dexamethasone)
- Fetal bovine serum
- Williams' E Medium, no phenol red
- Nunclon Sphera 96-well U-bottom plates
Please note that components are shipped separately due to differences in storage temperatures.
Simplified protocol
Cryopreserved cells can be thawed and plated at your convenience, in as little sa 60 minutes. Try it on your own, or request a demo with an experienced scientist.
Stringent quality control
Our high standards help ensure only the best spheroid-forming lots become available, maximizing your chance of success. In addition, each lot is tested for:
- Albumin production in monolayer and spheroid culture
- Phase I enzyme induction
- Bile duct formation (CFDA staining)
- Proper morphology
- Viability >75%
- Attachment efficiency
- Monolayer formation and integrity
Important! For best results, do not attempt hepatic spheroid formation in low attachment plates from other sources. Hepatocyte lots are qualified in Nunclon Sphera plates only and use of other plates can lead to variability in spheroid formation.
Flexible readouts
Analyze single spheroids, or pool several for detection in assays such as gene expression, immunostaining, viability, or CYP activity.
Specifications
Specifications
| Content And Storage | • Spheroid-qualified hepatocytes (1 mL, liquid N2 vapor phase only) |
| Cell Type | Hepatocytes |
| Form | Cryopreserved |
| Species | Human |
| Donor Source | Single Donor |
| Product Type | Hepatocytes |
| Quantity | 1 Kit |
| Shipping Condition | Cells: Dry Shipper, Plates: Room Temperature, Media/Reagents: Dry Ice |
Frequently Asked Questions (FAQs)
Our hepatocytes are prequalified and sold according to application, such as induction, short-term metabolism, and transporter uptake. If you require hepatocytes with particular P450 values or donor specifications, contact our Hepatic Biology Tech Support Team (hepaticproducts@thermofisher.com.com) or (866) 952-3559 to help with specific lot recommendations.
Thermo Fisher Scientific has a dedicated team to answer questions regarding hepatocyte cell culture. You can send a question by email (hepaticproducts@thermofisher.com) or contact us by phone: US toll-free: (866) 952-3559
Thermofisher Scientific works with a variety of human tissue sources, including Tissue and Organ Procurement Organizations, qualified Research Tissue Organizations and prominent academic and medical centers through collaborations that follow rigorous regulations, certifications and/or accreditations (Source Facilities). Tissues obtained through source facilities are consistent with the applicable legal and ethical practices of the United States.
Source Facilities are required by federal law to maintain accurate records and donor confidentiality as well as adhere to certain applicable laws governing tissue used for research globally. These include, but are not limited to:
- 45 CFR Part 46, subparts A, B, C, D where applicable (US federal guidance on the use of human subjects in research)
- Health Insurance Portability and Accountability Act (HIPAA) (addressing issues of confidentiality of personal information)
- National Organ Transplant Act (42 CFR Part 482)
- The Uniform Anatomical Gift Act (UAGA) of 1968, revised 1984
The human liver cells are derived from tissue obtained from accredited institutions. Consent is obtained by these institutions from the donor or the donor’s legal next of kin, for the use of the tissue and its derivatives for research purpose.
We have used a ratio between 10:1 and 3:1 for 3D-spheroid hepatocytes to stellate cells for a successful 3D co-culture. We would recommend testing different ratios as the cells may vary from one type of model to another.
For Research Use Only. Not for use in diagnostic procedures.