Learn More
Thermo Scientific™ HEPA Main Filter, for 3110 Series, 310 Series (w/ HEPA option), Steri-Cycle Series incubators, HEPA Main Filter; For 3110 Series, 310 Series (w/ HEPA option), Steri-Cycle Series incubators
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
Supplier:  Thermo Scientific™ 760175
Catalog No. 15467190
Only null left
Description
Description
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Product Suggestions
Product Suggestions