Learn More
BINDER™ HEPA Air Filter, Classification H14
Catalog No. 08101232
Change view
Click to view available options
Certifications/Compliance:
EN 1822-1:2009
Catalog No. Certifications/Compliance
08101232 EN 1822-1:2009
1 options
Catalog No. 08101232 Supplier BINDER™ Supplier No. 80122220
Fresh air supply for the Series FP Avantgarde.Line Drying and Heating Chambers.
  • Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
Certifications/Compliance EN 1822-1:2009
Description Removes >99.995% (at 0.3 μm)
For Use With (Equipment) BINDER Series FP Avantgarde.Line Drying and Heating Chambers
Type Air Filter
HEPA Filter H14