missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heneicosanoic Acid 98.0+%, TCI America™
Supplier: TCI America H001010G
Specifications
| Heneicosanoic Acid | |
| 75°C | |
| 171°C | |
| MFCD00002805 | |
| heneicosanoic acid, n-heneicosanoic acid, heneicosylic acid, henicosanoate, heneicosanate, heneicosanoate, n-heneicosanoate, heneicosanoicacid, heneicosanic acid, n-heneicosylic acid | |
| CCCCCCCCCCCCCCCCCCCCC(=O)O | |
| 326.565 | |
| CHEBI:39248 | |
| ≥98.0% (GC,T) |
| 2363-71-5 | |
| White | |
| C21H42O2 | |
| 10 g | |
| CKDDRHZIAZRDBW-UHFFFAOYSA-N | |
| henicosanoic acid | |
| 16898 | |
| 326.57 | |
| Crystalline Powder |
Chemical Identifiers
| 2363-71-5 | |
| 326.565 | |
| CKDDRHZIAZRDBW-UHFFFAOYSA-N | |
| 16898 | |
| henicosanoic acid |
| C21H42O2 | |
| MFCD00002805 | |
| heneicosanoic acid, n-heneicosanoic acid, heneicosylic acid, henicosanoate, heneicosanate, heneicosanoate, n-heneicosanoate, heneicosanoicacid, heneicosanic acid, n-heneicosylic acid | |
| CHEBI:39248 | |
| CCCCCCCCCCCCCCCCCCCCC(=O)O |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : No