missing translation for 'onlineSavingsMsg'
Learn More

Heneicosanoic Acid 98.0+%, TCI America™

Catalog No. H00101G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00101G 1 g
H001010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H00101G Supplier TCI America Supplier No. H00101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2363-71-5
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.565
MDL Number MFCD00002805
InChI Key CKDDRHZIAZRDBW-UHFFFAOYSA-N
Synonym heneicosanoic acid, n-heneicosanoic acid, heneicosylic acid, henicosanoate, heneicosanate, heneicosanoate, n-heneicosanoate, heneicosanoicacid, heneicosanic acid, n-heneicosylic acid
PubChem CID 16898
ChEBI CHEBI:39248
IUPAC Name henicosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 75°C
Color White
Boiling Point 171°C
Quantity 1 g
Formula Weight 326.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Heneicosanoic Acid
EINECSNumber (2)-0608
TSCA No