missing translation for 'onlineSavingsMsg'
Learn More

Heneicosane 99.0+%, TCI America™

Catalog No. H03671G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03671G 1 g
H036725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03671G Supplier TCI America Supplier No. H03671G

Chemical Identifiers

CAS 629-94-7
Molecular Formula C21H44
Molecular Weight (g/mol) 296.583
MDL Number MFCD00009346
InChI Key FNAZRRHPUDJQCJ-UHFFFAOYSA-N
Synonym heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
PubChem CID 12403
ChEBI CHEBI:32931
IUPAC Name henicosane
SMILES CCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 41°C
Color White
Boiling Point 356°C
Quantity 1 g
Formula Weight 296.58
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heneicosane
EINECSNumber (2)-0010
TSCA Yes