missing translation for 'onlineSavingsMsg'
Learn More

Heneicosafluoroundecanoic Acid 97.0+%, TCI America™

Catalog No. H12341G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12341G Supplier TCI America Supplier No. H12341G
Only null left

Chemical Identifiers

CAS 2058-94-8
Molecular Formula C11HF21O2
Molecular Weight (g/mol) 564.09
MDL Number MFCD00153268
InChI Key SIDINRCMMRKXGQ-UHFFFAOYSA-N
Synonym Perfluoroundecanoic Acid
PubChem CID 77222
ChEBI CHEBI:83493
IUPAC Name henicosafluoroundecanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 101°C
Color White
Boiling Point 160°C
Quantity 1 g
Formula Weight 564.09
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Heneicosafluoroundecanoic Acid
EINECSNumber (2)-1182&(2)-2659&(2)-2658
TSCA No