Learn More
Eurotrol Hemotrol
Supplier: Eurotrol 171.003.002
Hemotrol is a hemostatic agent used to achieve hemostasis in surgical procedures. It is composed of a plant-based chitosan derivative that activates platelets and the coagulation cascade upon contact with blood. The product comes in multiple formats including pads, powders, and solutions for versatility across applications. Hemotrol is biocompatible, absorbable, and easy to apply during surgery. It can be used in open and laparoscopic procedures across specialties including cardiovascular, orthopedic, general, plastic, and trauma surgery. The website indicates Hemotrol is effective at sealing bleeding tissues and assisting with coagulation.