Learn More
Bioassay Systems HEMOGLOBIN ASSAY REAGENT DFS Item
Supplier:  Bioassay Systems DIHB250REAGANTONLY
The Reagent for BioAssay Systems' Hemoglobin Assay Kit (Catalog# DIHB-250). For quantitative determination of hemoglobin and evaluation of drug effects on hemoglobin metabolism. Safe. Reagents are non-toxic. Linear detection range 0.9 - 200 mg /dL hemoglobin. The "mix-and-read" 5 minute procedure. Can be readily automated to process thousands of samples per day. Assays can be executed in 96-well plate or cuvet. Method: OD400 nm. Samples: Blood, serum, plasma, urine etc. A standard or calibrator is needed to run hemoglobin assays (see DIHB-250 assay kit).
Catalog No. NC0067379
Only null left
Explore available promotions