missing translation for 'onlineSavingsMsg'
Learn More

Hemin, 95%, MP Biomedicals™

Catalog No. p-4405879
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19882001 1 g
ICN19882010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19882001 Supplier MP Biomedicals Inc Supplier No. 0219882001
Edge

Chemical Identifiers

CAS 16009-13-5
Molecular Formula C34H32ClFeN4O4
Molecular Weight (g/mol) 651.95
MDL Number MFCD00010726
InChI Key BTIJJDXEELBZFS-HXFTUNQESA-K
Synonym chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin
PubChem CID 131675604
IUPAC Name 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
SMILES CC1=C(CCC(O)=O)\C2=C\C3=C(CCC(O)=O)C(C)=C4\C=C5/N=C(/C=C6\N([Fe](Cl)N34)\C(=C/C1=N2)C(C=C)=C6C)C(C=C)=C5C

Specifications

Color Black
Density 7.87g/cm3 at 25°C
Quantity 1 g
Formula Weight 652
Percent Purity 0.95
Physical Form Crystalline Powder
Chemical Name or Material Hemin