missing translation for 'onlineSavingsMsg'
Learn More

Hemimellitic Acid 98.0+%, TCI America™

$700.89 - $700.89

Chemical Identifiers

CAS 569-51-7
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.141
MDL Number MFCD00002468
InChI Key UJMDYLWCYJJYMO-UHFFFAOYSA-N
Synonym 1,2,3-Benzenetricarboxylic Acid
PubChem CID 11288
IUPAC Name benzene-1,2,3-tricarboxylic acid
SMILES C1=CC(=C(C(=C1)C(=O)O)C(=O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15925G
View Documents
TCI America
H15925G
5 g
Each for $700.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

569-51-7
210.141
UJMDYLWCYJJYMO-UHFFFAOYSA-N
11288
C1=CC(=C(C(=C1)C(=O)O)C(=O)O)C(=O)O
C9H6O6
MFCD00002468
1,2,3-Benzenetricarboxylic Acid
benzene-1,2,3-tricarboxylic acid

Specifications

569-51-7
White
MFCD00002468
1,2,3-Benzenetricarboxylic Acid
C1=CC(=C(C(=C1)C(=O)O)C(=O)O)C(=O)O
210.141
210.14
Crystalline Powder
194°C
C9H6O6
5 g
UJMDYLWCYJJYMO-UHFFFAOYSA-N
benzene-1,2,3-tricarboxylic acid
11288
≥98.0% (T)
Hemimellitic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1279

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'