missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin Stain Solution, Gill 3 Formulation, Triple Strength, Ricca Chemical

For Frozen or Plastic Imbedded Sections, and For More Intense Nuclei in Progressive Histology

$222.44 - $414.81

Specifications

Chemical Name or Material Hematoxylin Stain Solution
Name Note Gill 3 Formulation, Triple Strength
CAS 7732-18-5,107-21-1,64-19-7,7784-31-8,517-28-2,7681-55-2
CAS Min % 70.0,19.47,4.41,3.7,0.42,0.04
CAS Max % 73.0,20.27,4.59,3.85,0.44,0.04
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3537-16
View Documents
Ricca Chemical Company
353716
500 mL Natural Poly Bottle
Each for $222.44
Only null left
 
3537-32
View Documents
Ricca Chemical Company
353732
1 L Natural Poly Bottle
Each for $414.81
Only null left
 
Description

Description

Specifications

Specifications

Hematoxylin Stain Solution
7732-18-5,107-21-1,64-19-7,7784-31-8,517-28-2,7681-55-2
73.0,20.27,4.59,3.85,0.44,0.04
2 to 4
Gill 3 Formulation, Triple Strength
70.0,19.47,4.41,3.7,0.42,0.04
Dark Purple
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'