missing translation for 'onlineSavingsMsg'
Learn More

HEMATOGNOST Fe™, Staining Kit, MilliporeSigma™

Catalog No. M1120840001
Change view
Click to view available options
Quantity:
4 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1120840001 4 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1120840001 Supplier Technidata Supplier No. 1.12084.0001

Specifications

Packaging Fibre case
Quantity 4 x 250 mL
UN Number UN 3316
Physical Form Liquid
Chemical Name or Material HEMATOGNOST Fe™, Staining Kit