Learn More
Strem Chemicals Inc CAS# 1317-60-8. 250g. Iron(III) oxide, red (Hematite) (99.8%-Fe). MFCD00011008
Catalog No. 5090114645
Encompass
CAS# 1317-60-8. 250g. Iron(III) oxide, red (Hematite) (99.8%-Fe). MFCD00011008. Molecular Weight: 159.69. Molecular Formula: Fe2O3. Color/form: red pwdr. Strem# 93-2617. http://www.strem.com/catalog/v/93-2617/
Catalog No. 5090114645 Supplier Strem Chemicals Inc Supplier No. 932617250G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CAS# 1317-60-8. 250g. Iron(III) oxide, red (Hematite) (99.8%-Fe). MFCD00011008. Molecular Weight: 159.69. Molecular Formula: Fe2O3. Color/form: red pwdr. Strem# 93-2617. http://www.strem.com/catalog/v/93-2617/