Learn More
Hemastix™ Reagent Strips
Catalog No. S69043
Change view
Click to view available options
Product Type:
Hemastix™ Reagent Strip
Catalog No. Product Type
S69043 Hemastix™ Reagent Strip
1 options
Catalog No. S69043 Supplier Customer Channels Group (Ccg) Supplier No. 2135
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Quick-and-easy test can be used right at the crime scene.
Determine if what is being photographed at the scene is actually blood spatter.
  • Detect the peroxide-like activity of hemoglobin in a substance
  • Bottle of 50 strips
Specifications
For Use With (Application) For used at crime scenes to determine if what is being photographed at the scene is actually blood spatter
Product Type Hemastix™ Reagent Strip