Learn More
StatLab™ HemaDiff Rapid Stain Set
Description
- Entire procedure takes 30 seconds as air dried specimens are fixed in Solution I, then dipped in Solutions II and III
- Ideal for FNA cytology, differential staining of blood smears, microbiological organisms, H. pylori, and bone marrow specimens
- Versatile and very economical
- Includes 500 mL of fixative, xanthene, and thiazine
Specifications
Specifications
| Format | Kit |
| Product Type | Stain Kit |