Learn More
StatLab™ Hema-Diff 2 Xanthene
Description
- Entire procedure takes 30 seconds
- Air dried specimens are fixed in Solution I, then dipped in Solutions II and III
- Can also be used as a special stain for Helicobacter pylori
Specifications
Specifications
| For Use With (Application) | Differential Staining |
| Quantity | 1 gal. |
| Product Type | Xanthene |