Learn More
MilliporeSigma™ Supelco™ Helium Purifier, Stainless Steel fittings, Swagelok (Nuts and Ferrules included)
Catalog No. p-7191357
Change view
Click to view available options
Diameter (Metric) Inner:
1/4 in.
1/8 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111002588 1/4 in.
111016183 1/8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111002588 Supplier MilliporeSigma™ Supelco™ Supplier No. 27601U
Only null left
This multiple bed trap removes hydrocarbons, moisture, oxygen, carbon monoxide, and carbon dioxide from helium gas streams.
The amount of highly effective, high capacity adsorbent material in each bed is optimized so that breakthrough of each contaminant is as close to simultaneous as possible. This avoids the costly replacement observed with other broad-spectrum traps that have exhausted their capacity for one contaminant, but still have capacity for other contaminants. This trap will easily purify 99.997% purity helium to a cumulative level of 100 ppb (hydrocarbons + moisture + oxygen + carbon monoxide + carbon dioxide). Specifications: Output Purity: 99.99999%. Output Efficiency (total): <100 ppb. Output Efficiency (hydrocarbons): <20 ppb (as methane). Output Efficiency (moisture): <10 ppb. Output Efficiency (oxygen): <2 ppb. Output Efficiency (carbon monoxide): <20 ppb. Output Efficiency (carbon dioxide): <1 ppm. Capacity (hydrocarbons): 30 g (as methane). Capacity (moisture): 46 g. Capacity (oxygen): 1200 mg. Max. Flow Rate: 8 L/min. Max. Pressure: 500 psi. Max. Temp.: 100 CDimensions for both purifiers16.5 in. (41.91 cm) long x 2 5/16 in. (6.3 cm) diameter
Specifications
Type GC Accessories and Adsorbents
Diameter (Metric) Inner 1/4 in.
Includes Nuts and ferrules
Material Stainless Steel