missing translation for 'onlineSavingsMsg'
Learn More

Helicin 98.0+%, TCI America™

Catalog No. H09085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09085G 5 g
H09081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09085G Supplier TCI America Supplier No. H09085G
Only null left

Chemical Identifiers

CAS 618-65-5
Molecular Formula C13H16O7
Molecular Weight (g/mol) 284.26
MDL Number MFCD00006589
InChI Key BGOFCVIGEYGEOF-UJPOAAIJSA-N
Synonym helicin, 2-beta-d-glucopyranosyloxy benzaldehyde, unii-6n4ti8p9r5, 2-formylphenyl beta-d-glucopyranoside, salicylaldehyde beta-d-glucopyranoside, helicine, salicylaldehyde b-d-glucoside, salicylaldehyde, a-d-glucoside, salicylaldehyde-beta-d-glucoside
PubChem CID 101799
ChEBI CHEBI:73235
IUPAC Name 2-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}benzaldehyde
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2C=O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 5 g
Formula Weight 284.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Helicin