missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Stirring Bars

Stir bars for use with various Heidolph magnetic stirrers.

$85.52 - $997.26

Specifications

Autoclavable Autoclavable
Color White
For Use With (Application) Application Stirring
View More Specs

Products 4
Catalog Number Mfr. No. Length (Metric) Shape Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Shape Price Quantity & Availability  
05403925
View Documents Promotion Details
Heidolph™
015880330
25 mm Elliptical
Pack of 10 Each for $997.26
Only null left
 
05403918
View Documents Promotion Details
Heidolph™
015880260
10 mm Cross
Pack of 40 Each for $821.97
Only null left
 
05403919
View Documents Promotion Details
Heidolph™
015880270CA
16.5 mm Cross
Pack of 20 Each for $937.44
Only null left
 
13900813
View Documents Promotion Details
Heidolph™
036300930
25, 40, 50 mm Cylinder
Each for $85.52
Only null left
 
Description

Description

Stirring bars recommended for use with an Heidolph Magnetic Stirrers or Heidolph Magnetic Stirring Hotplates.

Variety of shapes to choose from

Specifications

Specifications

Autoclavable
White
Application Stirring
Videos
SDS
Documents

Documents