Learn More
Heidolph™ Radleys Jacketed Vessel Kit
Radleys jacketed vessel kit for use with Reactor-Ready™ and Reactor-Ready™ Duo lab reactors
Supplier:  Heidolph™ 015950105
 View more versions of this product
Catalog No. 05405257
Only null left
Description
Description
  • These cost effective vessel kits include the vessel and accessories you need to get started.
  • The Pt100 probe and anchor stirrer included in each kit are matched to the vessel size.
  • Each vessel kit includes reaction vessel, PTFE anchor stirrer, side arm couplings, Pt100 PTFE temperature probe with LEMO connector, and PTFE temperature probe adapter.
Specifications
Chemical Identifiers
446°F
Jacketed Vessel Kit
Reactor-Ready™ and Reactor-Ready™ Duo Lab Reactors
Specifications
446°F
500 mL
Reactor-Ready™ and Reactor-Ready™ Duo Lab Reactors
Single jacketed
Jacketed Vessel Kit
Product Suggestions
Product Suggestions