missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Radleys GreenHouse Plus Reaction Station Accessories: Reaction Block

Catalog No. 05403791
Change view
Click to view available options
Type:
Reaction Block
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05403791 Reaction Block
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05403791 Supplier Heidolph™ Supplier No. 015860620

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For GreenHouse Plus Reaction Stations

Specifications

For Use With (Equipment) Heidolph Radleys GreenHouse Plus Reaction Station™ and Blowdown Evaporator™
Type Reaction Block