Learn More
Heidolph™ Radleys GreenHouse Blowdown Evaporator™ Accessories: Blowdown Pin Plates
For use with GreenHouse Blowdown Evaporator™
$1606.09 - $1606.09
Specifications
| For Use With (Equipment) | Heidolph Radleys GreenHouse Blowdown Evaporator |
|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
05-403-794
|
Heidolph™
015860650 |
Long Blowdown Pin Plate, 6 x 4 |
Each for $1,606.09
|
|
|||||
Description
Interchangeable pin plates with either 8 or 24 hollow blowdown pins deliver an equal flow of gas to each tube, vial or well
Specifications
| Heidolph Radleys GreenHouse Blowdown Evaporator |