Learn More
Heidolph™ Radleys Carousel Reaction Station Accessory, Quick–Thread Glass Reaction Tubes
For use with Radleys Carousel 6 Plus™ and Carousel 12 Plus™ Reaction Stations
Supplier:  Heidolph™ 015862550
 View more versions of this product
Catalog No. 13900167
Only null left
Description
Description
150 mm x 24 mm ø tubes manufactured from heavy walled borosilicate glass feature a unique quickthread for fast attachment to the caps. Available in 6/pack and 12/pack
Specifications
Chemical Identifiers
Glass
Radleys Carousel 6 Plus™ and Carousel 12 Plus™ reaction system
20 mL
150 mm
24 mm
Quick Thread Glass Reaction Tube
0.68 oz.
5.9 in.
0.94 in.
Specifications
Glass
Radleys Carousel 6 Plus™ and Carousel 12 Plus™ reaction system
0.68 oz.
5.9 in.
0.94 in.
Quick Thread Glass Reaction Tube
12/Pk.
20 mL
150 mm
24 mm
Product Suggestions
Product Suggestions