Learn More
Heidolph™ Hei-FLOW Peristaltic Pump Accessory - Multi-Channel Pump Heads
Used to configure Heidolph Hei-FLOW Core 120, Hei-FLOW Expert 120 and Hei-FLOW Ultimate 120 Peristaltic Pumps for Multi-Channel use
Supplier:  Heidolph™ 036150480
 View more versions of this product
Catalog No. 13881147
Only null left
Description
Description
Increases throughput by running up to 12 individual cassettes on one single pump drive and pump head system.
  • Allows separate metering into multiple vessels with different feed rates in a single operation by using different tubing dimensions to adjust flow rates
  • Saves time by minimizing tube change efforts
  • Choice of standard 4-roller pump heads or 8-roller (for low pulsation)
  • Cassettes can be changed during operation
Specifications
Chemical Identifiers
Hei-FLOW Core 120, Hei-FLOW Expert 120 and Hei-FLOW Ultimate 120
Pump Head
Specifications
0.02 to 37 mL/min
4
Pump Head
Hei-FLOW Core 120, Hei-FLOW Expert 120 and Hei-FLOW Ultimate 120
8
Product Suggestions
Product Suggestions