Learn More
Heidolph™ Rotary Evaporator Accessories - Interchangeable Glassware
Catalog No. 13878569
Change view
Click to view available options
Type:
Clamp S 35
G1 Diagonal Condenser
G3 Vertical Condenser
Support for G5 Condenser (with condensate trap)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13878569 G1 Diagonal Condenser
13878578 Clamp S 35
13878585 G3 Vertical Condenser
13878635 Support for G5 Condenser (with condensate trap)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13878569 Supplier Heidolph™ Supplier No. 036301120
Includes
G1 Diagonal Condenser only
Five condenser assemblies allow customization of evaporator glassware for any application
  • G1 Diagonal Condenser
  • All glassware can be interchanged among rotary evaporator models
  • All sets are available with standard or plastic safety-coated glassware (halar coating for G5B and abcite coating for G1B/G3B/G6B)
Specifications
Type G1 Diagonal Condenser
For Use With (Equipment) Rotary Evaporator