Learn More
Heidolph™ Hei-Vap Rotary Evaporator Vapor Tubes
Catalog No. 13878568
Change view
Click to view available options
Taper:
24/40
29/32
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Taper
13878568 24/40
13878567 29/32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13878568 Supplier Heidolph™ Supplier No. 036301110
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Vapor tube only. (Clamping sleeve is sold separately.)
Accessory parts and components for Heidolph Hei-VAP Rotary Evaporators
  • The patented clamping sleeve system allows the removal of the vapor tube easily from the drive
  • Years of performance life reduce your spare parts budget up to 75%
  • The Hei-VAP vapor tube does not get stuck in the drive, thus eliminating the risk of broken glass
  • Immediate removal of the vapor tube from the drive facilitates your work
Specifications
Type Vapor Tube
For Use With (Equipment) Hei-VAP Rotary Evaporator
Includes Vapor tube only. (Clamping sleeve is sold separately; Heidolph No. 036308030.)
Taper 24/40