Learn More
Heidolph™ Hei-Vap Rotary Evaporator Vapor Tubes
Accessory parts and components for Heidolph Hei-VAP Rotary Evaporators
Supplier: Heidolph™ 036301100
Includes: Vapor tube only. (Clamping sleeve is sold separately.)
Description
- The patented clamping sleeve system allows the removal of the vapor tube easily from the drive
- Years of performance life reduce your spare parts budget up to 75%
- The Hei-VAP vapor tube does not get stuck in the drive, thus eliminating the risk of broken glass
- Immediate removal of the vapor tube from the drive facilitates your work
Chemical Identifiers
| Vapor Tube | |
| Hei-VAP Rotary Evaporator | |
| Vapor tube only. (Clamping sleeve is sold separately; Heidolph No. 036308030.) |
Specifications
| Vapor Tube | |
| Hei-VAP Rotary Evaporator | |
| Vapor tube only. (Clamping sleeve is sold separately; Heidolph No. 036308030.) | |
| 29/32 |