missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Guard Shield
Description
Guard Shield
- Strong and transparent guard shield for your protection while changing flasks
- Attached to the edge of the bath
Specifications
Specifications
| Color | Transparent |
| Type | Guard Shield |
| For Use With (Equipment) | Hei-VAP rotary evaporator |