Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Foam Brakes
Catalog No. 13878704
Change view
Click to view available options
Taper:
24/40
Catalog No. Taper
13878704 24/40
1 options
Catalog No. 13878704 Supplier Heidolph™ Supplier No. 036302470
For use with Heidolph Hei-VAP series rotary evaporators
  • The foam brake prevents contents of the evaporating flask from being drawn into the condenser in the event of bumping.
Specifications
Type Foam Brake
For Use With (Equipment) Hei-VAP Rotary Evaporator
Includes Flask clamp
Taper 24/40