Learn More
Heidolph™ Glassware - Wide Neck Flasks
Catalog No. 13900151
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
Includes:
2x B14/23 sidearms
2x B24/29 sidearms
B14/23 sidearms
B24/29 sidearm
With coupling
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Includes
13900151 50 mL B14/23 sidearms
13900153 100 mL 2x B14/23 sidearms
13900155 250 mL 2x B24/29 sidearms
13900152 100 mL B14/23 sidearms
13900154 250 mL B24/29 sidearm
13900150 250 mL With coupling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13900151 Supplier Heidolph™ Supplier No. 015861973
Flasks for use with Carousel 6 Plus Reaction Station
  • Available with sidearm, septaport and coupling
Specifications
Description Seal material: Silicone
Capacity (Metric) 50 mL
Material Glass
Includes B14/23 sidearms
Neck Style Wide
Standard Taper Joint B14/23 Side