Learn More
Heidolph™ Glassware - G3B Condenser
Catalog No. 13878586
Change view
Click to view available options
For Use With (Equipment):
Laborota Interchangeable Glassware
Catalog No. For Use With (Equipment)
13878586 Laborota Interchangeable Glassware
1 options
Catalog No. 13878586 Supplier Heidolph™ Supplier No. 036301290
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vertical condenser for all standard distillations; the option with the smallest space requirement
  • Transparent plastic coating (Surlyn-coated) for added safety
Specifications
Type G3B Vertical Condenser
For Use With (Equipment) Laborota Interchangeable Glassware
Material (Coating) Safety