Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Receiving Flasks
Description
Standard and Surlyn-coated receiving flasks for Heidolph Hei-VAP Rotary Evaporator
- NS 35/20 joint size
Specifications
Specifications
| Type | Coated Receiving Flask |
| For Use With (Equipment) | Hei-VAP Rotary Evaporator |
| Taper | 35/20 |
| Volume (Metric) | 2000 mL |