Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Receiving Flasks
Catalog No. 13878714
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Type:
Coated Receiving Flask
Standard Receiving Flask
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Type
13878714 2000 mL Standard Receiving Flask
13878707 250 mL Standard Receiving Flask
13878575 1000 mL Coated Receiving Flask
13878706 100 mL Coated Receiving Flask
13878716 3000 mL Standard Receiving Flask
13878710 500 mL Coated Receiving Flask
13878715 2000 mL Coated Receiving Flask
13878705 100 mL Standard Receiving Flask
13878574 1000 mL Standard Receiving Flask
13878717 3000 mL Coated Receiving Flask
13878708 250 mL Coated Receiving Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 13878714 Supplier Heidolph™ Supplier No. 036302570
Only null left
Coated or standard receiving flasks
Standard and Surlyn-coated receiving flasks for Heidolph Hei-VAP Rotary Evaporator
  • NS 35/20 joint size
Specifications
Type Standard Receiving Flask
For Use With (Equipment) Hei-VAP Rotary Evaporator
Taper 35/20
Volume (Metric) 2000 mL