Learn More
Heidolph™ Hei-Vap Rotary Evaporator Accessories - Glassware Sets
Catalog No. 13878600
Change view
Click to view available options
Type:
Glassware G1 Set
Glassware G1B Set
Glassware G3B Set
Taper:
NS 24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Taper
13878600 Glassware G3B Set NS 24/40
13878580 Glassware G1 Set NS 24/40
13878582 Glassware G1B Set NS 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13878600 Supplier Heidolph™ Supplier No. 036301430
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessories for Hei-VAP rotary evaporator
  • Glassware set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All glassware sets feature GL 10 thread; the inlet tube can be connected with a threaded fitting
  • All glassware sets are also available with transparent plastic halar coating for G5B and abcite coating for G1B/G3B/G6B
  • Glassware sets G3 and G6 allow for the automated distillation program AUTOaccurate
Specifications
Type Glassware G3B Set
For Use With (Equipment) Hei-VAP Rotary Evaporator
Includes Vertical condenser, 1000 mL NS 24/40 Joint sized evaporating flask, 1000 mL receiving flask, transparent plastic coating
Taper NS 24/40