Learn More
Heidolph™ Hei-VAP Industrial and Laborota 20 Rotary Evaporator Accessories
Catalog No. 13878862
Change view
Click to view available options
Type:
Tube Set
Vapor Temperature Sensor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13878862 Tube Set
13878860 Vapor Temperature Sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13878862 Supplier Heidolph™ Supplier No. 036303100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessories for use with Heidolph Hei-VAP Industrial and Laborota 20 rotary evaporators only
  • The vacuum sensor controls and displays the actual vacuum controls in addition the maximum permissible pressure.
  • The vacuum valve controls vacuum.
  • Vapor temperature sensor measure the temperature of vapors in the condenser.
  • Tubing set is 6 meter of vacuum and 10 meter of enforced cooling tube including hose clamps.
Specifications
Type Tube Set
For Use With (Equipment) Hei-VAP Industrial and Laborota 20 Rotary Evaporator