missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-SHAKE Stand Foot for Attachments

Catalog No. 12100026
Change view
Click to view available options
Material:
Electro-polished Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
12100026 Electro-polished Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12100026 Supplier Heidolph™ Supplier No. 036213733
Only null left

Base to secure Hei-SHAKE attachments to platform.

Base for attaching the racks for various vessels (test tubes, micro reaction vessels, etc.) to the platform of the selected platform shaker.

  • Micro Reaction Vessel Attachments - 12-100-027 and 13-100-028
  • Centrifuge Tube Attachments- 12-100-030 and 12-100-031
  • Test Tube Attachments- 12-100-034, 12-100-029 and 12-100-035
  • Tool-free assembly.
  • Material: electro-polished stainless steel
  • Characteristics: autoclavable and dishwasher safe

Specifications

Product Type Adapter
Material Electro-polished Stainless Steel
Color Silver
For Use With (Equipment) Hei-SHAKE Orbital Core platform shaker