missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-SHAKE Separatory Funnel Holder

Catalog No. 13687220
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
13687220 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13687220 Supplier Heidolph™ Supplier No. 036213741

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Separatory funnel attachment for Hei-SHAKE Orbital Core platform.

A flexible holder allows shaking separating funnels with a volume of 50 mL - 2 L. The single separating funnel holder is delivered in a pre-assembled way and you do not need tools for mounting it on a perforated platform with universal perforation being placed on the shaker's platform.

250 mL separatory funnel - maximum three vessels on the perforated platform 1000 and four vessels on the perforated platform 2000

2 L separatory funnel - maximum one vessel on the perforated platform 1000 and two vessels on the perforated platform 2000

Suitable for platform shakers with orbital or reciprocating shaking movement.

Specifications

Autoclavable Autoclavable
Color Silver
Holds Separatory Funnels
Material Polypropylene, Stainless Steel
Positions 1
Sterility Non-sterile
For Use With (Equipment) Separatory funnels 50 mL - 2 L
Product Type Funnel Holder