missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-SHAKE Micro Reaction Vessel Attachment with 36 Positions

Catalog No. 12100027
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
12100027 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12100027 Supplier Heidolph™ Supplier No. 036213734

Micro reaction vessel attachment for Hei-SHAKE Orbital Core platform shaker.

Attachment or rack for holding 36 micro reaction vessels such as Eppis 0.5 mL.

  • Material: stainless steel (frame) / polyoxymethylene (inner part)
  • Characteristics: autoclavable up to 121°C and dishwasher safe; angle adjustment up to 90° attachment without foot part can be used as table frame

This attachment is an optional accessory, must be ordered separately with the base 12-100-026.

Specifications

Autoclavable Autoclavable
Color Silver
Holds Micro Reaction Vessels
Material Stainless Steel (Frame), POM (Inner)
Positions 36
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Hei-SHAKE Orbital Core platform shaker
Product Type Vessel Rack
Shelves 1
Array 12 x 3
Show More Show Less