missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Hei-PLATE Mix 'n' Heat Magnetic Stirring Hotplate Accessory, Protective Cover
Description
- Replaceable cover against splashes and dripping water
Specifications
Specifications
| Color | Silver/White |
| For Use With (Equipment) | Hei-PLATE Mix 'n' Heat |
| Type | Silicone Protective Cover |
| Material | Silicone |