missing translation for 'onlineSavingsMsg'
Learn More

Hederagenin 98.0+%, TCI America™

Catalog No. p-7131140
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H164550MG 50 mg
H1645200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H164550MG Supplier TCI America Supplier No. H164550MG

Chemical Identifiers

CAS 465-99-6
Molecular Formula C30H48O4
Molecular Weight (g/mol) 472.71
MDL Number MFCD00017385
InChI Key PGOYMURMZNDHNS-MYPRUECHSA-N
Synonym 3beta,4alpha-3,23-Dihydroxyolean-12-en-28-oic Acid
PubChem CID 73299
ChEBI CHEBI:69579
IUPAC Name (4aS,6aR,6aS,6bR,8aR,9R,10S,12aR,14bS)-10-hydroxy-9-(hydroxymethyl)-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)CO)O)C)C)C2C1)C)C(=O)O)C

Specifications

Melting Point 334°C
Color White
Quantity 50 mg
Formula Weight 472.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hederagenin
TSCA No
Recommended Storage Refrigerator