missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hecogenin 80.0+%, TCI America™
Specifications
Specifications
| Color | Red-White |
| Quantity | 1 g |
| Formula Weight | 430.63 |
| Percent Purity | ≥80.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Hecogenin |
Safety and Handling
| TSCA | No |