missing translation for 'onlineSavingsMsg'
Learn More

Hecogenin 80.0+%, TCI America™

Catalog No. h0003100mg
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0003100MG 100 mg
H00031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0003100MG Supplier TCI America Supplier No. H0003100MG

Chemical Identifiers

CAS 467-55-0
Molecular Formula C27H42O4
Molecular Weight (g/mol) 430.63
MDL Number MFCD00067285
InChI Key QOLRLLFJMZLYQJ-LOBDNJQFSA-N
PubChem CID 91453
ChEBI CHEBI:5633
IUPAC Name (1'R,2R,2'S,4'S,5R,7'S,8'R,9'S,12'S,13'S,16'S,18'S)-16'-hydroxy-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-10'-one
SMILES C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC[C@H]5C[C@@H](O)CC[C@]5(C)[C@H]4CC(=O)[C@]23C)O[C@]11CC[C@@H](C)CO1

Specifications

Color Red-White
Quantity 100 mg
Formula Weight 430.63
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hecogenin