missing translation for 'onlineSavingsMsg'
Learn More

Hecogenin Acetate 90.0+%, TCI America™

Catalog No. H05371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05371G 1 g
H05375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05371G Supplier TCI America Supplier No. H05371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 915-35-5
Molecular Formula C29H44O5
Molecular Weight (g/mol) 472.67
MDL Number MFCD00005088
InChI Key CVKZWRTYHCDWTE-UHFFFAOYNA-N
PubChem CID 101906
IUPAC Name 5,7',9',13'-tetramethyl-10'-oxo-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-16'-yl acetate
SMILES CC1C2C(CC3C4CCC5CC(CCC5(C)C4CC(=O)C23C)OC(C)=O)OC11CCC(C)CO1

Specifications

Color White
Quantity 1 g
Formula Weight 472.67
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hecogenin Acetate