Learn More
Synthware™ Heavy Wall Pressure Vessels
GREENER_CHOICE
Vessels feature an internal thread for use with Teflon™ bushing as a pressure seal. Unique design allows the vessel to be loosened and tightened by hand for easy access.
Supplier:  Synthware™ P160704
 View more versions of this product
Catalog No. 31502377
Only null left
Description
Description
  • Internal thread for use with Teflon™ bushing as a pressure seal
  • Vessel can be loosened/tightened by hand for easy access
  • O-ring is positioned on the bushing above the thread for improved sealing
  • Complete apparatus includes vessel, Teflon™ bushing, and EP™ O-ring
  • Ideal for pressure reactions and applications such as polymerization and halogen analysis by thermal decomposition in the presence of lime
Specifications
Chemical Identifiers
Borosilicate Glass
Pressure Reactions, Polymerization, and Halogen Analysis
Vessel, Teflon™ Bushing, EP™ O-Ring
Cylindrical
Specifications
Borosilicate Glass
Pressure Reactions, Polymerization, and Halogen Analysis
80 mm
13 mm
Vessel, Teflon™ Bushing, EP™ O-Ring
Cylindrical
4 mL
#7
Product Suggestions
Product Suggestions