missing translation for 'onlineSavingsMsg'
Learn More

SPEX CertiPrep™ Heavy Metals Testing Kit

Catalog No. SPXHMKIT
Change view
Click to view available options
Volume (Metric):
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
SPXHMKIT 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXHMKIT Supplier SPEX CertiPrep™ Supplier No. HMKIT
Only null left

Heavy Metals Testing Kit, 1,000 ug/mL, for ICP-M

Claritas PPT Grade Heavy Metals Testing Kit for ICP-MS, 1,000 ug/mL (1,000 ppm), 6 each 30mL

Kit contains:

  • Arsenic, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
  • Cadmium, 1,000μg/mL, for ICP-MS, 30mL,Matrix: 2% HNO3
  • Chromium, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3
  • Mercury, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
  • Lead, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3

Specifications

Volume (Metric) 30 mL
Storage Requirements Ambient
Type Certified Reference Material