Learn More
Keystone Cleanroom Products Heavy Duty Sterilization Wraps
These wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications. For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
$643.76 - $2,427.89
Specifications
| Basis Weight | 50 gsm |
|---|---|
| Autoclavable | Autoclavable |
| Certifications/Compliance | CoC |
| Closure Type | None |
| Color | Light Blue |
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
|
17010627
|
Keystone Cleanroom Products
KCRP13000808 |
8 in. | 8 in. |
Case of 500 Each for $735.72
|
|
||||
|
17010626
|
Keystone Cleanroom Products
KCRP13000909 |
9 in. | 9 in. |
Case of 500 Each for $809.30
|
|
||||
|
17010625
|
Keystone Cleanroom Products
KCRP13000912 |
12 in. | 9 in. |
Case of 500 Each for $993.23
|
|
||||
|
17010624
|
Keystone Cleanroom Products
KCRP13001212 |
12 in. | 12 in. |
Case of 500 Each for $938.05
|
|
||||
|
17010623
|
Keystone Cleanroom Products
KCRP13001515 |
15 in. | 15 in. |
Case of 500 Each for $1,195.55
|
|
||||
|
17010622
|
Keystone Cleanroom Products
KCRP13001818 |
18 in. | 18 in. |
Case of 500 Each for $1,655.39
|
|
||||
|
17010621
|
Keystone Cleanroom Products
KCRP13002020 |
20 in. | 20 in. |
Case of 500 Each for $1,912.90
|
|
||||
|
17010620
|
Keystone Cleanroom Products
KCRP13002424 |
24 in. | 24 in. |
Case of 125 Each for $643.76
|
|
||||
|
17010619
|
Keystone Cleanroom Products
KCRP13003030 |
30 in. | 30 in. |
Case of 125 Each for $1,048.41
|
|
||||
|
17010618
|
Keystone Cleanroom Products
KCRP13003636 |
36 in. | 36 in. |
Case of 125 Each for $1,434.66
|
|
||||
|
17010617
|
Keystone Cleanroom Products
KCRP13004040 |
40 in. | 40 in. |
Case of 125 Each for $1,839.31
|
|
||||
|
17010616
|
Keystone Cleanroom Products
KCRP13004545 |
45 in. | 45 in. |
Case of 125 Each for $2,170.40
|
|
||||
|
17010615
|
Keystone Cleanroom Products
KCRP13004548 |
48 in. | 45 in. |
Case of 125 Each for $2,427.89
|
|
||||
|
17010614
|
Keystone Cleanroom Products
KCRP13004848 |
48 in. | 48 in. |
Case of 125 Each for $2,427.89
|
|
||||
|
17010613
|
Keystone Cleanroom Products
KCRP13005454 |
54 in. | 54 in. |
Case of 50 Each for $1,030.01
|
|
||||
|
17010612
|
Keystone Cleanroom Products
KCRP13005472 |
72 in. | 54 in. |
Case of 50 Each for $1,600.21
|
|
||||
|
17010611
|
Keystone Cleanroom Products
KCRP13005490 |
90 in. | 54 in. |
Case of 25 Each for $1,085.19
|
|
||||
|
17010610
|
Keystone Cleanroom Products
KCRP13006060 |
60 in. | 60 in. |
Case of 50 Each for $1,692.17
|
|
||||
|
17010609
|
Keystone Cleanroom Products
KCRP13006080 |
80 in. | 60 in. |
Case of 25 Each for $901.26
|
|
||||
Description
- Manufactured and packaged in a certified ISO8 Cleanroom
- 2-Ply
- Clean, no cellulose
- Strong, rip and tear resistant 50 gsm SMS material
- An extensive range of sizes available means less cutting
- Hydrophobic barrier allows sterilization gasses, but prevents liquid and microbial penetration
- Sub packed into sealed labeled poly bags with lot number and date of manufacture for complete traceability
- Certificate of Conformance sent with each shipment
- Made in the USA
sterilization, sterilization sheets, sterilization wraps, autoclavable wraps
Specifications
| 50 gsm | |
| CoC | |
| Light Blue | |
| Steam Sterilization | |
| Non-sterile | |
| 50 gsm | |
| 50 gsm |
| Autoclavable | |
| None | |
| Single-use | |
| Non-woven Fabric | |
| Yes | |
| Sterilization Wrap (Heavy-duty) |