missing translation for 'onlineSavingsMsg'
Learn More

CHECKERS™ Heavy Duty Spring Mount for Non-Lighted Whips

Catalog No. 19085961
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19085961 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19085961 Supplier CHECKERS™ Supplier No. FS7006DUD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Heavy Duty Spring Mount for Non-Lighted Whips, and for use with models in 3, 5, 6, and 8 foot lengths

Specifications

Material Chrome
Packaging Quantity 1 Ea.
For Use With (Equipment) Non-lighted whips
Color Silver