missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Heavy-Duty SMS Coverall

Catalog No. 19068497
Change view
Click to view available options
Size:
X-Large
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19068497 X-Large
19055366 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19068497 Supplier Keystone Safety Supplier No. CVLSMSREGEXLWHITE
Only null left

Keystone Safety Heavy-Duty SMS Coverall is a latex free coverall that is made from high quality materials and comfortable to wear. It conforms to ANSI specs and has a strong/high tensile strength. This durable yet light-weight coverall is disposable with surged seams.

  • Fluid and Tear/Puncture Resistant
  • 100% Latex Free
  • Soft & Durable

Specifications

Product Type Single-use Coveralls
Size X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Particle, Liquid
Latex Free Yes
No. of Pockets None
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less