Learn More
Keystone Cleanroom Products™ Heavy Duty SMS Autoclavable Bowl Cover
This 50 g/m2 SMS Autoclavable Bowl Cover provides a microbial barrier ideal for pharmaceutical production applications.
$1904.67 - $8419.75
Specifications
Autoclavable Autoclavable
Closure Type Elastic
Color Light Blue
Disposable Single-use
Material Reinforced Multicoated Plastic
View More Specs
Products 10
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
17-200-291
View Documents
Keystone Cleanroom Products™
BWLKCRP30020
11 to 16 in. 11 to 16 in.
Case of 500 Each for $5,803.03
Only null left
17-200-290
View Documents
Keystone Cleanroom Products™
BWLKCRP30024
12 to 20 in. 12 to 20 in.
Case of 500 Each for $5,981.04
Only null left
17-200-289
View Documents
Keystone Cleanroom Products™
BWLKCRP30030
20 to 26 in. 20 to 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17-200-288
View Documents
Keystone Cleanroom Products™
BWLKCRP30036
26 to 30 in. 26 to 30 in.
Case of 100 Each for $3,186.32
Only null left
17-200-287
View Documents
Keystone Cleanroom Products™
BWLKCRP30040
27 to 34 in. 27 to 34 in.
Case of 50 Each for $1,904.67
Only null left
17-200-286
View Documents
Keystone Cleanroom Products™
BWLKCRP30042
28 to 35 in. 28 to 35 in.
Case of 50 Each for $2,527.71
Only null left
17-200-285
View Documents
Keystone Cleanroom Products™
BWLKCRP30048
30 to 40 in. 30 to 40 in.
Case of 50 Each for $2,937.12
Only null left
17-200-284
View Documents
Keystone Cleanroom Products™
BWLKCRP30050
31 to 41 in. 31 to 41 in.
Case of 50 Each for $2,990.53
Only null left
17-200-283
View Documents
Keystone Cleanroom Products™
BWLKCRP30054
37 to 47 in. 37 to 47 in.
Case of 50 Each for $3,738.14
Only null left
17-200-282
View Documents
Keystone Cleanroom Products™
BWLKCRP30060
40 to 50 in. 40 to 50 in.
Case of 50 Each for $3,791.54
Only null left
Description
Description
  • Save time by using the stopper bowl cover instead of wrapping with sheets of SMS material and autoclave tape
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • After the outer bag is removed, this stopper bowl cover should remain in place, providing protection from microbial and particulate contamination while the remaining set-up activities are performed
  • Manufactured and packaged in a Certified ISO 8 cleanroom
  • Designed for autoclave cycles (121°C to 135°C)
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Hydrophobic and resists tearing and punctures
  • Lint-free
  • Conveniently packaged: 1 cover per heat-sealed and labeled poly bag
  • Certificates of Conformance sent with all shipments
  • Made in USA
Specifications
Specifications
Autoclavable
Light Blue
Reinforced Multicoated Plastic
121°C to 135°C
Elastic
Single-use
Non-sterile
Round Cover
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'